missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hexafluorophosphate, TRC

Product Code. 30334662
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334662 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334662 Supplier Toronto Research Chemicals Supplier No. TRCA63402525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Ammonium Hexafluorophosphate
CAS 16941-11-0
Quantity 25 g
Molecular Formula F6H4NP
Synonym Hexafluoro-phosphate(1-) Ammonium, Ammonium Hexafluorophosphate, Ammonium Hexafluorophosphate(1-), NSC 403554, NSC 407930
InChI Formula InChI=1S/F6P.H3N/c1-7(2,3,4,5)6;/h;1H3/q-1;/p+1
SMILES [NH4+].F[P-](F)(F)(F)(F)F
IUPAC Name azanium;hexafluorophosphate
Molecular Weight (g/mol) 163
Formula Weight 162.9986
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.