missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hexafluorophosphate, TRC

Product Code. 30339436
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339436 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339436 Supplier Toronto Research Chemicals Supplier No. TRCA6340255G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Hexafluorophosphate
CAS 16941-11-0
Quantity 5 g
Molecular Formula F6H4NP
Synonym Hexafluoro-phosphate(1-) Ammonium, Ammonium Hexafluorophosphate, Ammonium Hexafluorophosphate(1-), NSC 403554, NSC 407930
InChI Formula InChI=1S/F6P.H3N/c1-7(2,3,4,5)6;/h;1H3/q-1;/p+1
SMILES [NH4+].F[P-](F)(F)(F)(F)F
IUPAC Name azanium;hexafluorophosphate
Molecular Weight (g/mol) 163
Formula Weight 162.9986
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.