missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hexafluorophosphate, Honeywell Fluka™

Product Code. 15640910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15640910 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15640910 Supplier Honeywell-Fluka Supplier No. 0982025G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥98.0% (as NH4PF6, gravimetric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 16941-11-0
Quantity 25 g
Molecular Formula F6H4NP
Linear Formula NH4PF6
MDL Number MFCD00064642
UN Number 3260
Synonym ammonium hexafluorophosphate, ammonium hexafluorophosphate v, ammonium hexafluophosphate, ammoniumhexafluorophosphate 6ci,7ci, unii-ns308031pj, azanium hexafluorophosphate, nh4pf6, ksc492g2j, ammonium hexafluoro-, e5-phosphanuide, ammonium phosphorus hexafluoride
Solubility Information Soluble in water , 50mg/mL
InChI Key NIZXKAYXSNUDOU-UHFFFAOYSA-O
SMILES [NH4+].F[P-](F)(F)(F)(F)F
IUPAC Name azanium;hexafluorophosphate
Molecular Weight (g/mol) 163.003
PubChem CID 9793912
Formula Weight 163g/mol
Percent Purity ≥98% (as NH4PF6)
Density 2.18g/mL at 25°C
Chemical Name or Material Ammonium Hexafluorophosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.