missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorophosphate, 99.5%

Product Code. 11337167
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337167 100 g 100g
11327167 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337167 Supplier Thermo Scientific Alfa Aesar Supplier No. 011495.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonium hexafluorophosphate, 99.5%, Alfa Aesar, Quantity: 100g, CAS: 16941-11-0, Melting Point: 198 deg.C, Molecular Formula: F6H4NP, Molecular Weight: 163.003 | CAS: 16941-11-0 | F6H4NP | 163.003 g/mol

Ammonium hexafluorophosphate is used as a preservative. It is used as a starting material in the preparation of other hexafluorophosphates. Further, it is used in synthesis of organometalic compounds, especially rhodicinium hexafluorophosphate, which has inert and non-coordinating counter ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16941-11-0
Molecular Formula F6H4NP
Molecular Weight (g/mol) 163.003
MDL Number MFCD00064642
InChI Key NIZXKAYXSNUDOU-UHFFFAOYSA-O
Synonym ammonium hexafluorophosphate, ammonium hexafluorophosphate v, ammonium hexafluophosphate, ammoniumhexafluorophosphate 6ci,7ci, unii-ns308031pj, azanium hexafluorophosphate, nh4pf6, ksc492g2j, ammonium hexafluoro-, e5-phosphanuide, ammonium phosphorus hexafluoride
PubChem CID 9793912
IUPAC Name azanium;hexafluorophosphate
SMILES [NH4+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point 198°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5%
Solubility Information Freely soluble in water (75g/100ml). Soluble in acetone,methanol,ethanol,methyl acetate.Soluble in water.
Formula Weight 163
Sensitivity Hygroscopic
Density 2.180 g/mL
Chemical Name or Material Ammonium hexafluorophosphate
compliance-icons
Product Identifier
  • Ammonium hexafluorophosphate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.