missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorogermanate, 99.99% min (metals basis)

Product Code. 11334868
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334868 5 g 5g
11344868 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334868 Supplier Thermo Scientific Alfa Aesar Supplier No. 043179.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16962-47-3 | H8F6GeN2

Ammonium hexafluorogermanate is used to substitutes Al with Ge in Y type zeolites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16962-47-3
Molecular Formula H8F6GeN2
MDL Number MFCD00010880

Specifications

Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula (NH4)2GeF6
UN Number UN3288
Solubility Information Soluble in water.
Formula Weight 222.71
Percent Purity ≥99.99%
Density 2.564 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexafluorogermanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.