missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachlorostannate(IV), Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11388668
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388668 50 g 50g
11368668 10 g 10g
11378668 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11388668 Supplier Thermo Scientific Alfa Aesar Supplier No. 040750.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16960-53-5
Molecular Formula Cl6H8N2Sn
Molecular Weight (g/mol) 367.488
MDL Number MFCD00036340
InChI Key UDMAVAHFQCVSSE-UHFFFAOYSA-J
Synonym ammonium tin chloride, stannic ammonium chloride, ammonium hexachlorostannate, diammonium hexachlorostannate, ammonium hexachlorostannate 2-, diammonium hexachlorostannate 2-, ammonium tin 4+ chloride 2:1:6, diazanium tin 4+ hexachloride, stannate 2-, hexachloro-, diammonium, oc-6-11, stannate 2-, hexachloro-, ammonium 1:2 , oc-6-11
PubChem CID 167595
IUPAC Name diazanium;tin(4+);hexachloride
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula (NH4)2SnCl6
Solubility Information Soluble in water
Formula Weight 367.48
Percent Purity 99.999%
Grade Puratronic
Density 2.4 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachlorostannate(IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.