missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachlororuthenate(IV), 99.99%, (trace metal basis)

Product Code. 11914371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11914371 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11914371 Supplier Thermo Scientific Acros Supplier No. 436930010
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18746-63-9 | Cl6H8N2Ru | 349.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 18746-63-9
Color Black to Brown
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99%
Molecular Formula Cl6H8N2Ru
Linear Formula (NH4)2RuCl6
MDL Number MFCD00015962
Synonym ammonium hexachlororuthenate iv, diammonium hexachlororuthenate, ammonium chlororuthenate, ammonium hexachlororuthenate, epitope id:151520, ruthenium iv-ammonium chloride, ammonium hexachloridoruthenate iv, ammonium hexachloridoruthenate 2-, diammonium hexachlororuthenate 2-
Solubility Information Solubility in water: soluble
InChI Key DHTSBQSPUBOKKI-UHFFFAOYSA-J
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+4]
IUPAC Name diazanium;hexachlororuthenium(2-)
Molecular Weight (g/mol) 349.85
PubChem CID 46173598
ChEBI CHEBI:60134
Formula Weight 349.85
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ammonium hexachlororuthenate(IV)
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium hexachlororuthenate(IV)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.