missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachlororhodate(III) hydrate, Premion™, 99.99% (metals basis)

Product Code. 11331858
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11331858

Brand: Thermo Scientific Alfa Aesar 011812.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15336-18-2 | Cl6H12N3Rh | 369.72 g/mol

Ammonium hexachlororhodate(III) hydrate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15336-18-2
Molecular Formula Cl6H12N3Rh
Molecular Weight (g/mol) 369.72
MDL Number MFCD00003394
InChI Key QNMMDVLCBKRUJU-UHFFFAOYSA-K
Synonym ammonium hexachlororhodate iii, triammonium hexachlororhodate, triammonium hexachlororhodate 3-, epitope id:151523, rhodium iii-ammonium chloride, ammonium hexachloridorhodate 3-, ammonium hexachloridorhodate iii, ammonium hexachlororhodate iii , reagent grade
PubChem CID 16211509
ChEBI CHEBI:60128
IUPAC Name triazanium;hexachlororhodium(3-)
SMILES [NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Color Red
Physical Form Powder
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 369.74 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.