missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroplatinate(IV), Pt 43.4% min

Product Code. 11367607
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367607 1 g 1g
11377607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367607 Supplier Thermo Scientific Alfa Aesar Supplier No. 012882.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Electroplating of platinum, manufacturing of platinum sponge | CAS: 16919-58-7 | Cl6H8N2Pt | 443.86 g/mol

Electroplating of platinum, manufacturing of platinum spongeAmmonium hexachloroplatinate(IV) is used in electroplating of platinum and to make spongy platinum. It reacts with copper(II) acetate and hydrogen sulfide to prepare Cu2PtIIPt3IVS8 which finds application in diamagnetic materials and semiconductors. It is also used in platinum plating. Further, it acts as an analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16919-58-7
Molecular Formula Cl6H8N2Pt
Molecular Weight (g/mol) 443.86
MDL Number MFCD00010886
InChI Key PCCGQTHFYHJATL-UHFFFAOYSA-J
Synonym ammonium hexachloroplatinate iv, ammonium chloroplatinate, ammonium hexachloroplatinate, ammonium platinic chloride, ammonium platinum chloride, diammonium hexachloroplatinate 2-, diammonium platinum hexachloride, ammoniumhexachloroplatinate, epitope id:151516, platinum iv-ammonium chloride
PubChem CID 16211460
ChEBI CHEBI:59604
IUPAC Name diazanium;hexachloroplatinum(2-)
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]

Specifications

Melting Point (decomposition)
Color Yellow
Physical Form Powder
Quantity 1 g
UN Number UN3288
Merck Index 14,548
Solubility Information Slightly soluble in water; Insoluble in alcohol,ether and concentrated hydrochloric acid.
Formula Weight 443.89
Concentration or Composition (by Analyte or Components) Pt 43.4% min.
Sensitivity Hygroscopic
Density 3.06 g/mL
Chemical Name or Material Ammonium hexachloroplatinate(IV)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.