missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroosmate(IV), Premion™, 99.99% (metals basis), Os 42.8% min

Product Code. 11346957
Change view
Click to view available options
Quantity:
0.5 g
1 g
5 g
Unit Size:
0.5g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346957 1 g 1g
11326957 0.5 g 0.5g
11336957 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346957 Supplier Thermo Scientific Alfa Aesar Supplier No. 010823.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12125-08-5 | Cl6H8N2Os | 439.00 g/mol

Ammonium hexachloroosmate(IV) is suitable for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-08-5
Molecular Formula Cl6H8N2Os
Molecular Weight (g/mol) 439.00
MDL Number MFCD00010883
InChI Key JRHMYOOMZKAPKT-UHFFFAOYSA-J
Synonym ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
PubChem CID 11729867
SMILES [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl

Specifikationer

Melting Point 170°C (sublimation)
Color Brown to Black
Physical Form Powder
Quantity 1 g
Sensitivity Hygroscopic
Density 2.9 g/mL
Linear Formula (NH4)2OsCl6
Merck Index 14,536
Solubility Information Soluble in water.
IUPAC Name diazanium;hexachloroosmium(2-)
Formula Weight 438.99
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Os 42.8% min.
Assay (metals basis)
Chemical Name or Material Ammonium hexachloroosmate(IV)
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Ammonium hexachloroosmiate(IV)
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Kompletterande information
  • MIXTURE LIST-Contain: Ammonium hexachloroosmiate(IV)

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.