missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroosmate(IV), 99.99%, (trace metal basis)

Product Code. 11914361 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11914361 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11914361 Supplier Thermo Scientific Acros Supplier No. 197630010
  • ONLINE EXCLUSIVE PRICE
    £149.00
    £128.65 / 1g
    Save £20.35
    13% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12125-08-5 | Cl6H8N2Os | 439.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 170.0°C
CAS 12125-08-5
Color Black to Red
Physical Form Powder
Quantity 1 g
Packaging Glass bottle
Assay Percent Range 99.99%
Molecular Formula Cl6H8N2Os
Linear Formula (NH4)2OsCl6
MDL Number MFCD00010883
Merck Index 15,533
Synonym ammonium hexachloroosmate iv, nh4 2oscl6, osmium iv-ammonium chloride, diazanium;hexachloroosmium 2-, diammonium hexachloroosmiumdiuide, ammonium hexachloroosmiate iv , premion
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
InChI Key JRHMYOOMZKAPKT-UHFFFAOYSA-J
SMILES [NH4+].[NH4+].Cl[Os+4](Cl)(Cl)(Cl)(Cl)Cl
IUPAC Name diammonium hexachloroosmiumtetrakis(ylium)
Molecular Weight (g/mol) 439.00
PubChem CID 11729867
Formula Weight 439.02
Percent Purity 99.99%
Grade Trace Metal Basis
Chemical Name or Material Ammonium hexachloroosmate(IV)
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium Hexachloroosmate(IV)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.