missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroiridate(IV), Premion™, 99.994% (metals basis), Ir 41% min

Product Code. 11310989
Click to view available options
Quantity:
0.1 g
1 g
5 g
Unit Size:
0.1g
1g
5g
This item is not returnable. View return policy

Product Code. 11310989

Brand: Thermo Scientific Alfa Aesar 010712.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16940-92-4 | Cl6H8IrN2 | 440.99 g/mol

It is a key intermediate in the isolation of iridium from ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-92-4
Molecular Formula Cl6H8IrN2
Molecular Weight (g/mol) 440.99
MDL Number MFCD00010881
InChI Key QOPXOJZBUJGPAG-UHFFFAOYSA-J
Synonym ammonium hexachloroiridate iv, ammonium chloroiridate, diammonium hexachloroiridate, diammonium hexachloroiridate 2-, ammonium hexachloroiridate, epitope id:151519, iridium iv-ammonium chloride, iridium iii-ammonium chloride, ammonium hexachloridoiridate iv, diazanium;hexachloroiridium 2-
PubChem CID 16211476
ChEBI CHEBI:60132
SMILES [NH4+].[NH4+].Cl[Ir+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Color Black-Red
Physical Form Crystalline Powder
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name diazanium;hexachloroiridium(2-)
Formula Weight 441.02
Percent Purity 99.994%
Grade Premion
Concentration or Composition (by Analyte or Components) Ir 41% min.
Sensitivity Hygroscopic
Density 2.86 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium hexachloroiridate(IV)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.