missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroiridate(III) hydrate, Thermo Scientific™

Product Code. 11387477
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387477 5 g 5g
11377477 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387477 Supplier Thermo Scientific Alfa Aesar Supplier No. 012632.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29796-57-4
Molecular Formula Cl6H14IrN3O
Molecular Weight (g/mol) 477.049
MDL Number MFCD00003393
InChI Key LJHBEBFYNMDGCZ-UHFFFAOYSA-K
Synonym ammonium hexachloroiridate iii hydrate, triammonium hexachloroiridiumtriuide hydrate
PubChem CID 18366487
IUPAC Name triazanium;hexachloroiridium(3-);hydrate
SMILES [NH4+].[NH4+].[NH4+].O.Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 459.06 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Ammonium hexachloroiridate(III) hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.