missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexachloroiridate(III) hydrate, Premion™, 99.99% (metals basis)

Product Code. 11340989
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11340989

Brand: Thermo Scientific Alfa Aesar 010713.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15752-05-3 | Cl6H12IrN3 | 459.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15752-05-3
Molecular Formula Cl6H12IrN3
Molecular Weight (g/mol) 459.03
MDL Number MFCD00003393
InChI Key AIDFGYMTQWWVES-UHFFFAOYSA-K
IUPAC Name iridium(3+) triammonium hexachloride
SMILES [NH4+].[NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ir+3]

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 459.06 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Ammonium hexachloroiridate(III) hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.