missing translation for 'onlineSavingsMsg'
Learn More

Ammonium glycyrrhizinate, MedChemExpress

Product Code. 16471969
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16471969 Supplier Medchem Express Supplier No. HY76225
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ammonium glycyrrhizinate (Monoammonium glycyrrhizinate) has various pharmacological actions such as anti-inflammatory, antiallergic, antigastriculcer, and antihepatitis activities.

Ammonium glycyrrhizinate (Monoammonium glycyrrhizinate) has various pharmacological actions such as anti-inflammatory, antiallergic, antigastriculcer, and antihepatitis activities.

Specifications

Chemical Name or Material Ammonium glycyrrhizinate
Physical Form Solid
CAS 53956-04-0
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C42H65NO16
For Use With (Application) COVID-19-immunoregulation
Synonym Monoammonium glycyrrhizinateGlycyrrhizic acid ammonium saltAmmonium glycyrrhizate
Solubility Information DMSO : 100 mg/mL (119.05 mM; Need ultrasonic)
SMILES O=C1[C@]([C@](CC2)(C)[C@@]3(C)C([C@]([C@@]4(C)CC3)([H])C[C@@](CC4)(C)C(O)=O)=C1)([H])[C@](CC5)(C)[C@]2([H])C([C@H]5O[C@@H](O[C@H](C(O)=O)[C@H]6O)[C@H](O[C@@H]([C@H](O)[C@@H](O)[C@@H]7O)O[C@@H]7C(O)=O)[C@H]6O)(C)C.N
Molecular Weight (g/mol) 839.96
Formula Weight 839.96
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.