missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Glycyrrhizinate (>80%), TRC

Product Code. 30136469
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136469 100 g 100g
30003773 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136469 Supplier Toronto Research Chemicals Supplier No. TRCA634010100G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53956-04-0
Molecular Formula C42H65NO16
Molecular Weight (g/mol) 839.96
Synonym (3β,20β)-20-Carboxy-11-oxo-30-norolean-12-en-3-yl 2-O-β-D-glucopyranuronosyl- α-D-glucopyranosiduronic Acid Ammonium Salt, (3β,20β)-20-Carboxy-11-oxo-30-norolean-12-en-3-yl-2-O-β-D-Glucopyranuronosyl-α-D-Glucopyranosiduronic Acid Monoammonium Salt, (+)-Glycyram, 18β-Glycyrrhizic acid Monoammonium Salt, Ammonium Glycyrrhizate, Ammonium Glycyrrhizinate, Glycamil, Glycymin, Glycyron Ammonium Salt, Glycyron monoammonium salt, Glycyrram, Glycyrrhizic Acid Ammonium Salt, Glycyrrhizic Acid Monoammonium Salt, Glycyrrhizin Ammonium Salt, Glycyrrhizin Monoammonium Salt, Magnasweet, Monoammonium 18β-Glycyrrhizinate, Monoammonium Glycyrrhizate, Monoammonium Glycyrrhizinate, NSC 2800, NSC 35348;
IUPAC Name (2S,3S,4S,5R,6R)-6-(((2S,3R,4S,5S,6S)-6-carboxy-2-(((3S,4aR,6aR,6bS,8aS,11S,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,14,14a,14b-icosahydropicen-3-yl)oxy)-4,5-dihydroxytetrahydro-2H-pyran-3-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
SMILES [H][C@@]12C[C@@H](C)(C(O)=O)CC[C@]1(C)CC[C@H]3(C)C2=CC([C@@H]4([H])[C@@]3(C)CC[C@H]5([H])[C@]4(C)CC[C@H](O[C@H]6O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]6O[C@H]7[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O7)C5(C)C)=O.N

Specifications

Quantity 100 g
InChI Formula InChI=1S/C42H62O16.H3N/c1-37(2)21-8-11-42(7)31(20(43)16-18-19-17-39(4,36(53)54)13-12-38(19,3)14-15-41(18,42)6)40(21,5)10-9-22(37)55-35-30(26(47)25(46)29(57-35)33(51)52)58-34-27(48)23(44)24(45)28(56-34)32(49)50;/h16,19,21-31,34-35,44-48H,8-15,17H2,1-7H3,(H,49,50)(H,51,52)(H,53,54);1H3/t19-,21-,22-,23-,24-,25-,26-,27+,28-,29-,30+,31+,34-,35-,38+,39-,40-,41+,42+;/m0./s1
Formula Weight 839.43
Chemical Name or Material Ammonium glycyrrhizate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.