missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, TRC

Product Code. 30005075
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005075 50 g 50g
30004125 100 g 100g
30003578 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005075 Supplier Toronto Research Chemicals Supplier No. TRCA63400550G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-69-2
Molecular Formula CH2O2 . NH3
Molecular Weight (g/mol) 63.06
Synonym Formic Acid Ammonium
IUPAC Name azanium;formate
SMILES N.OC=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3
Chemical Name or Material Ammonium Formate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.