missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Formate, Reagent grade, 97%, Honeywell Burdick & Jackson™
Reagent Grade, 97%
£10.76 - £175.41
Chemical Identifiers
CAS | 540-69-2 |
---|---|
Molecular Formula | CH5NO2 |
Molecular Weight (g/mol) | 63.056 |
MDL Number | MFCD00013103 |
InChI Key | VZTDIZULWFCMLS-UHFFFAOYSA-N |
Synonym | ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny |
PubChem CID | 2723923 |
ChEBI | CHEBI:63050 |
IUPAC Name | azanium;formate |
SMILES | C(=O)[O-].[NH4+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15641500
|
Honeywell Research Chemicals
156264-100G |
100 g | Poly Bottle | ||||||
15671500
|
Honeywell Research Chemicals
156264-500G |
500 g | Poly Bottle | ||||||
15651500
|
Honeywell Research Chemicals
156264-1KG |
1 kg | Poly Bottle | ||||||
15661500
|
Honeywell Research Chemicals
156264-2.5KG |
2.5 kg | Poly Bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
540-69-2 | |
63.056 | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
2723923 | |
azanium;formate |
CH5NO2 | |
MFCD00013103 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
CHEBI:63050 | |
C(=O)[O-].[NH4+] |
Specifications
116°C | |
White | |
CH5NO2 | |
MFCD00013103 | |
3625095 | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
azanium;formate | |
2723923 | |
63.06g/mol | |
Reagent | |
1.26g/mL at 25°C |
540-69-2 | |
6.0 to 7.0 (at 20°C) | |
HCO2NH4 | |
NONH for all modes of transport | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
C(=O)[O-].[NH4+] | |
63.056 | |
CHEBI:63050 | |
97% | |
Crystals/Granules | |
Ammonium Formate |
Safety and Handling
missing translation for 'einecsNumber' : 208-753-9
missing translation for 'rtecsNumber' : BQ6650000