missing translation for 'onlineSavingsMsg'
Learn More

Ammonium formate, Certified AR for analysis, Fisher Chemical

Product Code. 10203230 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10203230 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10203230 Supplier Fisher Chemical Supplier No. A/5080/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 115°C
CAS 540-69-2
Color White
pH 6.0 to 7.0
Physical Form Solid
Quantity 500 g
Molecular Formula CH5NO2
MDL Number 13103
InChI Key VZTDIZULWFCMLS-UHFFFAOYSA-N
SMILES N.OC=O
IUPAC Name formic acid amine
Molecular Weight (g/mol) 63.06
Formula Weight 63.06g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium formate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.