missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, Puriss. p.a., ACS Reagent, ≥98%, Solstice

Product Code. 15692970 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
50 kg
6 x 1 kg
Unit Size:
100g
1kg
50kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15692970 6 x 1 kg Pack of 6
15662970 100 g 100g
15672970 1 kg 1kg
15682970 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15692970 Supplier Honeywell-Fluka Supplier No. 301016X1KG
  • ONLINE EXCLUSIVE PRICE
    £796.00
    £733.53 / Pack of 6
    Save £62.47
    7% Off
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Quantity 6 x 1 kg
Linear Formula NH4F
UN Number UN2505
Formula Weight 37.04
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Residue after Ignition ≤0.005% (as SO4)
Chemical Name or Material Ammonium Fluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.