missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, ACS, 98.0% min

Product Code. 11387297
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 x 1 kg
Unit Size:
1kg
250g
25g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387297 1 kg 1kg
11397297 25 g 25g
11377297 250 g 250g
11367297 5 x 1 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11387297 Supplier Thermo Scientific Alfa Aesar Supplier No. 011568.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12125-01-8 | FH4N | 37.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color White
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 98% min.
Solubility Information Freely soluble in water. Slightly soluble in alcohol.
Formula Weight 37.04
Grade ACS
Sensitivity Hygroscopic
Appearance White crystalline
Density 1.01
Chemical Name or Material Ammonium fluoride, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.