Learn More
Ammonium fluoride, 98+%, for analysis, Thermo Scientific Chemicals
£16.00 - £124.00
Chemical Identifiers
CAS | 12125-01-8 |
---|---|
Molecular Formula | H4FN |
Molecular Weight (g/mol) | 37.04 |
MDL Number | MFCD00011423 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10666314
|
Thermo Scientific Acros
393190500 |
50 g | Plastic bottle |
£16.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10065353
|
Thermo Scientific Acros
393192500 |
250 g | Plastic bottle |
£47.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10126863
|
Thermo Scientific Acros
393190010 |
1 kg | Plastic bottle |
£124.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12125-01-8 | |
H4FN | |
37.04 | |
MFCD00011423 |
Specifications
12125-01-8 | |
Liquid or Solid | |
H4FN | |
MFCD00011423 | |
Solubility in water: soluble. Other solubilities: slightly soluble in alcohol | |
37.04 | |
Analytical | |
Ammonium fluoride |
Colorless to Yellow or White to Yellow | |
98+% | |
NH4F | |
15, 519 | |
37.04 | |
≥98% | |
0.82 |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 235-185-9
- Ammonium fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only