missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 98+%, ACS reagent

Product Code. 10095573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10095573 2.5 kg Plastic bottle 2.5kg
10546792 5 g Plastic bottle 5g
10742022 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10095573 Supplier Thermo Scientific Acros Supplier No. 423290025
  • ONLINE EXCLUSIVE PRICE
    £257.00
    £249.00 / 2.5kg
    Save £8.00
    3% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

CAS: 12125-01-8 | FH4N | 37.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98+%
Linear Formula NH4F
Merck Index 15, 519
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 37.04
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.01% max
Chemical Name or Material Ammonium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.