missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, 0.5M solution in methanol, AcroSeal™, Thermo Scientific™

Product Code. 10098319 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10098319 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10098319 Supplier Thermo Scientific Acros Supplier No. 428891000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 65.0°C
Melting Point -98.0°C
CAS 12125-01-8, 67-56-1
Color Colorless
Physical Form Liquid
Quantity 100 mL
Molecular Formula FH4N
Linear Formula NH4F
MDL Number MFCD00011423
Flash Point 11°C
Merck Index 15, 519
Solubility Information Solubility in water: soluble
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
SMILES N.F
IUPAC Name amine hydrofluoride
Molecular Weight (g/mol) 37.04
Formula Weight 37.04
Concentration or Composition (by Analyte or Components) 0.45 to 0.55M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.82
Density 0.8200g/mL
Chemical Name or Material Ammonium fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium fluoride, 0.5M solution in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.