missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Ferric Disulfate Dodecahydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Ammonium iron(III) disulfate dodecahydrate |
| CAS | 7783-83-7 |
| Molecular Formula | Fe . H4 N . 12 H2 O . 2 O4 S |
| Quantity | 25 g |
| Synonym | Sulfuric acid, ammonium iron(3+) salt (2:1:1), dodecahydrate (8CI,9CI), Ammonium ferric disulfate dodecahydrate, Ammonium ferric sulfate dodecahydrate, Ammonium iron disulfate dodecahydrate, Ammonium iron sulfate ((NH4)Fe(SO4)2) dodecahydrate, Ammonium ir |
| InChI Formula | InChI=1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3 |
| SMILES | [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-] |
| IUPAC Name | azanium;iron(3+);disulfate;dodecahydrate |
| Molecular Weight (g/mol) | 482.192 |
| Formula Weight | 482 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?