missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Ferric Disulfate Dodecahydrate, TRC

Product Code. 30336540
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30336540 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30336540

Brand: Toronto Research Chemicals TRCA634113100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium iron(III) disulfate dodecahydrate
CAS 7783-83-7
Molecular Formula Fe . H4 N . 12 H2 O . 2 O4 S
Quantity 100 g
Synonym Sulfuric acid, ammonium iron(3+) salt (2:1:1), dodecahydrate (8CI,9CI), Ammonium ferric disulfate dodecahydrate, Ammonium ferric sulfate dodecahydrate, Ammonium iron disulfate dodecahydrate, Ammonium iron sulfate ((NH4)Fe(SO4)2) dodecahydrate, Ammonium ir
InChI Formula InChI=1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-]
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
Molecular Weight (g/mol) 482.192
Formula Weight 482
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.