missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, 98+%, ACS reagent

Product Code. p-45457 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10475851

Brand: Thermo Scientific Acros 423385000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190.0°C
Color White
pH 3.8 to 4.4 (5% soln. at 25°C)
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range 98+%
Linear Formula NH4H2PO4
Merck Index 15, 541
Solubility Information Solubility in water: 368g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in acetone
IUPAC Name azanium;dihydrogen phosphate
Formula Weight 115.03
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Density 1.8
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.