missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen phosphate, Puratronic™, 99.995% (metals basis)

Product Code. 11300829
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11300829 100 g 100g
11310829 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11300829 Supplier Thermo Scientific Alfa Aesar Supplier No. 010633.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol

Crystals of this compound are used in transducers as an alternative to Quartz and Rochelle salt. Monoammonium phosphate is one of the components of agricultural fertilizers. It supplies elements nitrogen and phosphorus to the soil. It is also a component of some dry chemical fire extinguishers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name azanium;dihydrogen phosphate
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190°C
Color White
Physical Form Crystalline
Quantity 100 g
Linear Formula NH4H2PO4
Merck Index 14,543
Solubility Information Soluble in water and ethanol. Insoluble in acetone
Formula Weight 115.03
Percent Purity 99.995%
Grade Puratronic
Odor Odorless
Density 1.803 g/mL
Assay (metals basis)
Chemical Name or Material Ammonium dihydrogen phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.