missing translation for 'onlineSavingsMsg'
Learn More

Ammonium dihydrogen arsenate, 98%

Product Code. 11383897
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383897 50 g 50g
11373897 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11383897 Supplier Thermo Scientific Alfa Aesar Supplier No. 031493.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 13462-93-6 | AsH6NO4 | 158.973 g/mol

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13462-93-6
Molecular Formula AsH6NO4
Molecular Weight (g/mol) 158.973
MDL Number MFCD00064864
InChI Key VTWJXWNBCACAAK-UHFFFAOYSA-N
Synonym ammonium dihydrogen arsenate, ammonium arsenate ortho, ammonium dihydrogenarsenate, unii-0kyl9q413a, arsenic acid h3aso4 , monoammonium salt, acmc-20alsv, azanium dihydrogen arsorate, arsenic acid h3aso4 ,ammonium salt 1:1, arsenic acid h3aso4 , ammonium salt 1:1
PubChem CID 166814
IUPAC Name azanium;dihydrogen arsorate
SMILES [NH4+].O[As](=O)(O)[O-]

Specifications

Melting Point 300°C (decomposition)
Physical Form Crystalline
Quantity 50 g
UN Number UN1557
Solubility Information Soluble in water. (337.4mg/ml at 0°C).
Formula Weight 158.98
Percent Purity 98%
Refractive Index 1.577
Density 2.311 g/mL
Chemical Name or Material Ammonium dihydrogen arsenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.