missing translation for 'onlineSavingsMsg'
Learn More

Ammonium diamminetetrathiocyanatochromate(III) monohydrate, ACS, 93.0% min

Product Code. 11315039
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11315039

Brand: Thermo Scientific Alfa Aesar 087777.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to precipitate primary and secondary amines as their ammonium salts | CAS: 13573-17-6 | C4H10CrN7S4·H2O

Ammonium diamminetetrathiocyanatochromate(III) monohydrate is used to precipitate primary and secondary amines as their ammonium salts. It is a color reagent and precipitant for mercury. It is also used as a precipitant for primary and secondary amines and certain amino acids. It is a reagent used for quantitative studies, including titrimetric, gravimetric, and spechtometric assays. Additionally, Reinecke salt has also been shown to determine the concentration of Dimenhydrinate using a spectrophotometric procedure. Other studies use Reinecke salt as a chemical actinometer to calculate the photodegradation of Manidipine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13573-17-6
Molecular Formula C4H10CrN7S4·H2O
MDL Number MFCD00066638
Synonym Reinecke salt; Ammonium reineckate

Specifications

Color Red
Physical Form Powder
Quantity 100 g
Linear Formula NH4[Cr(NH3)2(SCN)4]·H2O
UN Number UN3077
Merck Index 14,8128
Solubility Information Insoluble in dilute hydrochloric acid; Moderately soluble in water; Soluble in alcohol
Formula Weight 354.44 (336.42 Anhydrous)
Percent Purity ≥93.0%
Grade ACS
Odor Odorless
Chemical Name or Material Ammonium diamminetetrathiocyanatochromate (III) monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.