missing translation for 'onlineSavingsMsg'
Learn More

Ammonium copper(II) chloride dihydrate

Product Code. 11343508
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11343508

Brand: Thermo Scientific Alfa Aesar 039127.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10060-13-6 | Cl4CuH12N2O2 | 277.454 g/mol

Ammonium copper(II) chloride dihydrate is used in the determination iron of content in steel. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10060-13-6
Molecular Formula Cl4CuH12N2O2
Molecular Weight (g/mol) 277.454
MDL Number MFCD00150384
InChI Key LLMDTWKBBWCWNN-UHFFFAOYSA-L
Synonym ammonium tetrachlorocuprate ii dihydrate, cupric ammonium chloride dihydrate, ammonium copper ii chloride dihydrate, diammonium tetrachlorocuprate ii dihydrate, ammonium chlorocuprate ii, copper ii ammonium chloride dihydrate, diammonium tetrachlorocuprate dihydrate, copper +2 diammonium chloride dihydrate, diammonium copper ii chloride dihydrate, ammonium tetrachlorocuprated ii dihydrate
PubChem CID 16210985
IUPAC Name diazanium;tetrachlorocopper(2-);dihydrate
SMILES [NH4+].[NH4+].O.O.Cl[Cu-2](Cl)(Cl)Cl

Specifications

Melting Point 110°C (decomposition)
Color Blue
Physical Form Crystals
Quantity 500 g
Merck Index 14,514
Solubility Information Soluble in water,acids and alcohol; Slightly soluble in ammonia.
Formula Weight 277.46 (241.43 Anhydrous)
Density 1.993 g/mL
Chemical Name or Material Ammonium copper(II) chloride dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.