missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cobalt(II) sulfate hexahydrate, 98%

Product Code. 11413344
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413344 100 g 100g
11403344 25 g 25g
11423344 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413344 Supplier Thermo Scientific Alfa Aesar Supplier No. A10627.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13586-38-4 | (NH4)2Co(SO4)2·6H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13586-38-4
Molecular Formula (NH4)2Co(SO4)2·6H2O
MDL Number MFCD00151049
Synonym Cobalt(II) ammonium sulfate; Cobaltous ammonium sulfate

Specifications

Melting Point ∼120°C (decomposition)
Quantity 100 g
UN Number UN3077
Formula Weight 395.21 (287.13 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 1.902
Chemical Name or Material Ammonium cobalt (II) sulfate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.