missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cobalt(II) phosphate, anhydrous, 98%

Product Code. 11439997
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11439997

Brand: Thermo Scientific Alfa Aesar B22934.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14590-13-7 | CoH4NO4P | 171.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14590-13-7
Molecular Formula CoH4NO4P
Molecular Weight (g/mol) 171.94
MDL Number MFCD02683100
InChI Key FCXHHGTWMFGYPP-UHFFFAOYSA-L
Synonym ammonium cobaltous phosphate, ammonium cobalt ii phosphate, ammonium cobalt phosphate, kobaltammoniumphosphat, acmc-20alt0, ammonium cobalt orthophosphate, azanium cobalt 2+ phosphate, phosphoric acid, ammonium cobalt 2+ salt 1:1:1, ammonium cobalt ii phosphate, anhydrous, phosphoric acid,ammonium cobalt 2+ salt 1:1:1
PubChem CID 167120
IUPAC Name azanium;cobalt(2+);phosphate
SMILES [NH4+].[Co++].[O-]P([O-])([O-])=O

Specifications

Quantity 250 g
Linear Formula NH4CoPO4
Merck Index 14,513
Formula Weight 171.94
Percent Purity 98%
Chemical Name or Material Ammonium cobalt(II) phosphate, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.