missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, Puratronic™, 99.999% (metals basis)

Product Code. 11370819
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11370819 250 g 250g
11390819 50 g 50g
11380819 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11370819 Supplier Thermo Scientific Alfa Aesar Supplier No. 010632.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12125-02-9 | ClH4N | 53.49 g/mol

Ammonium chloride finds its use as a flux in preparing metals for galvanizing, soldering or tin coating. While mainly referred as a nitrogen source fertilizer, it is also used in cleaning tip of soldering iron, hair shampoo as a thickening agent, some batteries as electrolyte, tanning, textile printing, dyeing, and cleaning products. In laboratories it is used to produce cooling baths and for producing buffers. Along with certain metals, it is used for effecting certain reductions in organic synthesis. It can be used as a precursor to ammonium cyanate, which is employed in the Wöhler synthesis of urea. It facilitates several organic transformations in a mild and efficient manner - for example in Ulmann coupling, cross-aldol condensation, Claisen rearrangement, reduction, thia-Michael addition and synthesis of several monocyclic, fused or spiro bicyclic nitrogen heterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340° (sublimation)
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 100.00%
Solubility Information Highly soluble in water. Soluble in liquid ammonia,acetone,hydrazine,alcohol. Insoluble in diethyl ether and ethyl acetate.
IUPAC Name azanium;chloride
Formula Weight 53.49
Odor Odorless
Refractive Index 1.642
Sensitivity Hygroscopic
Appearance White powder
Density 1.53
Chemical Name or Material Ammonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.