missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10213900 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
2kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10213900 2 kg Plastic powder jar 2kg
10060030 500 g Plastic powder jar 500g
10151750 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10213900 Supplier Fisher Chemical Supplier No. A/3920/61
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

H4ClN, CAS Number-12125-02-9, 5kg, White, 53.49g/mol, 340 deg.C, 11420, Plastic keg, Solid, 1.3mbar 160 | CAS: 12125-02-9 | ClH4N | 53.49 g/mol

H4ClN, CAS Number-12125-02-9, 5kg, White, 53.49g/mol, 340 deg.C, 11420, Plastic keg, Solid, 1.3mbar 160

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Melting Point 340°C
Color White
pH 5
Physical Form Solid
Quantity 2 kg
Packaging Plastic powder jar
Vapor Pressure 1.3mbar 160
Formula Weight 53.49g/mol
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.