missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, 99.7%, MP Biomedicals™

Product Code. 11436502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. Quantity unitSize
11436502 1 kg 1kg
1 options
This item is not returnable. View return policy

Product Code. 11436502

Brand: MP Biomedicals 0215010991

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 12125-02-9
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 0.997
Molecular Formula ClH4N
MDL Number MFCD00011420
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILES N.Cl
IUPAC Name amine hydrochloride
Molecular Weight (g/mol) 53.49
PubChem CID 25517
ChEBI CHEBI:31206
Formula Weight 53.5
Chemical Name or Material Ammonium chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.