missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, 98+%

Product Code. 11447227
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447227 5000 g 5000g
11437227 250 g 250g
11314609 500 g 500g
11427227 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11447227 Supplier Thermo Scientific Alfa Aesar Supplier No. A15000.0I
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12125-02-9 | ClH4N | 53.49 g/mol

Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium Chloride is commonly used as a buffer solution. It is used in the manufacture of fireworks, safety explosives and smokes. It is used to control pH levels in chemicals, to produce sodium carbonate, as a fertilizer, in electronics and in metallurgy. Also used as a flux for cleaning, as a flavoring agent, as an expectorant in cough medicine and as a systemic acidifying agent. It is widely used as an electrolyte in zinc-carbon batteries. It has application in leather industries for dyeing, tanning, textile printing and to luster cotton.

Solubility
Soluble in water, methanol, liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate.

Notes
Hygroscopic. Incompatible with silver nitrate and strong oxidizers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340°C (sublimation)
Quantity 5000 g
Linear Formula NH4Cl
Beilstein 4371014
Merck Index 14,509
Solubility Information Soluble in water,methanol,liquid ammonia and acetone. Insoluble in diethyl ether and ethyl acetate.
IUPAC Name azanium;chloride
Formula Weight 53.49
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.642
Sensitivity Hygroscopic
Density 1.53
Chemical Name or Material Ammonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Ammonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.