missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Ammonium Chloride
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Description
Ammonium chloride, an inorganic compound, is a white crystalline salt, highly soluble in water.
Ammonium chloride, an inorganic compound, is a white crystalline salt, highly soluble in water.
Specifications
Specifications
| Chemical Name or Material | Ammonium Chloride |
| CAS | 12125-02-9 |
| Quantity | 500 g |
| Molecular Formula | NH4Cl |
| Grade | ACS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction