missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cerium(IV) sulfate dihydrate, 95%, ACS reagent

Product Code. 11984371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11984371 2.5 kg Glass Bottle 2.5kg
10410351 100 g Glass Bottle 100g
10125520 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11984371 Supplier Thermo Scientific Acros Supplier No. 325790025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10378-47-9 | H16CeN4O16S4·2H2O | 632.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10378-47-9
Molecular Formula H16CeN4O16S4·2H2O
Molecular Weight (g/mol) 632.54
Synonym Ceric ammonium sulfate

Specifications

Melting Point 130.0°C
Color Orange
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 95%
Linear Formula (NH4)4Ce(SO4)4·2H2O
Fieser 08,8
Solubility Information Solubility in water: hydrolysis. Other solubilities: insoluble in ethanol
Formula Weight 632.54
Percent Purity 95%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Ammonium cerium(IV) sulfate dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ammonium cerium(IV) sulfate dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.