missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(IV) Nitrate, TRC

Product Code. 30339643
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30339643 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30339643

Brand: Toronto Research Chemicals TRCA63393025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium cerium nitrate
CAS 16774-21-3
Molecular Formula Ce . 4 N O3 . 2 H N O3 . 2 H3 N
Quantity 25 g
Synonym CAN, Ceric ammonium nitrate, Diammonium cerium(IV) nitrate
InChI Formula InChI=1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;
SMILES N.N.[Ce+4].O[N+](=O)[O-].O[N+](=O)[O-].[O-][N+](=O)[O-].[O-][N+](=O)[O-].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
IUPAC Name azane;cerium(4+);nitric acid;tetranitrate
Molecular Weight (g/mol) 548.22
Formula Weight 547.9011
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.