missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cerium(IV) nitrate, Purum p.a., ≥98.0% (T), Honeywell™

Product Code. 16685763 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16685763 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16685763 Supplier Honeywell Research Chemicals Supplier No. 222505KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 214°C (Decomposition)
CAS 16774-21-3
Color Orange
CAS Max % ≤ 100%
Quantity 5 kg
Molecular Formula CeH8N8O18
Linear Formula Ce(NH4)2(NO3)6
Synonym Ceric ammonium nitrate
InChI Key WIBGOERAEYJBOT-UHFFFAOYSA-N
SMILES N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular Weight (g/mol) 548.22
Grade Reagent
Density 2.49 g/cm3 (20°C)
Chemical Name or Material Ammonium cerium(IV) nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.