missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(Iv) Nitrate, ≥98.0% (T), purum p.a., Solstice

Product Code. 15642190 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15642190 100 g Poly Bottle 100g
15652190 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15642190 Supplier Honeywell Research Chemicals Supplier No. 22250100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
MDL Number MFCD00151121

Specifications

Quantity 100 g
Linear Formula Ce(NH4)2(NO3)6
UN Number UN3085
Formula Weight 548.22g/mol
Percent Purity ≥98.0% (T)
Grade purum p.a.
Packaging Poly Bottle
Chemical Name or Material Ammonium Cerium(Iv) Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.