missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, MP Biomedicals™

Product Code. 11416502 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416502 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11416502 Supplier MP Biomedicals Supplier No. 0215010880
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 10361-29-2
Color White
Physical Form Crystalline Powder
Quantity 100 g
Molecular Formula CH8N2O3
Synonym hartshorn salt, bis soda triamine, ammonium carbonate, acs, bis carbonic acid triamine
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]
IUPAC Name diazanium;carbonate
Molecular Weight (g/mol) 96.086
PubChem CID 517111
Formula Weight 96.09
Chemical Name or Material Ammonium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.