missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, Honeywell™ Fluka™

Product Code. p-7120167 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15687670 100 g Plastic Bottle 100g
15607680 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15687670 Supplier Honeywell-Fluka Supplier No. A9516100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purity (Titration by NaOH)≥30.0 %

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]

Specifications

Color White
pH 9 (at 100g/L 20°C)
Physical Form Solid
Quantity 100 g
Linear Formula (NH4)2CO3
Beilstein 4253487
Formula Weight 96.09g/mol
Odor Ammonia stinging
Packaging Plastic Bottle
Density 0.9g/cm3 at 20°C; Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.