missing translation for 'onlineSavingsMsg'
Learn More

Ammonium carbonate, Puratronic™, 99.999% (metals basis)

Product Code. p-4407215
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11312078

Brand: Thermo Scientific Alfa Aesar 012980.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol

Ammonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient in some cough syrups. It is the main component in smelling salts and an active ingredient in some smokeless tobacco products, shampoos and dyes used in textile industries. It is an important predecessor to the modern leavening agent’s baking soda and baking powder. It serves as a foaming agent for the production of expanded material. It is also used as a reagent for analytical purposes in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
MDL Number MFCD00010890

Specifications

Melting Point 60° (sublimation)
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.999% (metals basis)
Molecular Formula C2H11N3O5
Beilstein 3627235
Synonym ammonium carbonate nf, ugam, ammonium bicarbonate-ammonium carbamate, ammonium carbonate chips nf, ammonium bicarbonate, mixt. with ammonium carbamate, ammonium carbamate, mixt. with ammonium bicarbonate, monoammonium carbonate mixture with ammonium carbamate, carbamic acid, monoammonium salt, mixt. with ammonium carbonate, diazanium hydrogen carbonate carbamate
Solubility Information Soluble in water.
InChI Key YNSUIDWLZCFLML-UHFFFAOYSA-N
SMILES C(=O)(N)[O-].C(=O)(O)[O-].[NH4+].[NH4+]
IUPAC Name diazanium;hydrogen carbonate;carbamate
Molecular Weight (g/mol) 157.126
PubChem CID 24660
Formula Weight 96.09
Sensitivity Hygroscopic
Chemical Name or Material Ammonium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.