missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, ≥30% NH3 basis (T), Honeywell™ Fluka™

Product Code. 15686950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15686950 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15686950 Supplier Honeywell-Fluka Supplier No. 74415250G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for HPLC,≥30% NH3 basis (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 506-87-6
Color White
pH 9 (at 100.0g/L 20°C)
Physical Form Solid
Quantity 250 g
Molecular Formula CH8N2O3
Linear Formula (NH4)2CO3
MDL Number MFCD00010890
UN Number UN3077
Beilstein 4253487
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]
IUPAC Name diazanium;carbonate
Molecular Weight (g/mol) 96.086
PubChem CID 517111
Formula Weight 96.09g/mol
Percent Purity ≥30% (NH3 basis (T))
Odor Ammonia stinging
Packaging Poly bottle
Density 0.9g/cu.cm at 20°C; Vapor Density: 2.7 (vs air)
Chemical Name or Material Ammonium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.