missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bromide, ACS, 99.0% min

Product Code. 11377837
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
Unit Size:
100g
25g
2kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377837 500 g 500g
11367837 25 g 25g
11357837 100 g 100g
11347837 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11377837 Supplier Thermo Scientific Alfa Aesar Supplier No. 014257.36
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 12124-97-9 | BrH4N | 97.94 g/mol

Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woodsAmmonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12124-97-9
Molecular Formula BrH4N
Molecular Weight (g/mol) 97.94
MDL Number MFCD00011419
InChI Key SWLVFNYSXGMGBS-UHFFFAOYSA-N
IUPAC Name ammonium bromide
SMILES [NH4+].[Br-]

Specifications

Melting Point 452° (sublimation)
Color Colorless
Physical Form Solid
Quantity 500 g
Sensitivity Hygroscopic
Assay Percent Range 99% min.
Density 2.429
Solubility Information Freely soluble in water,methanol,ethanol,and acetone. Slightly soluble in ether. Practically insoluble in ethyl acetateSoluble in water,methanol,ethanol and acetone. Slightly soluble in ether.
Formula Weight 97.97
Grade ACS
Chemical Name or Material Ammonium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.