missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bisulfite - In water, TRC

Product Code. 30335244
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 30335244

Brand: Toronto Research Chemicals TRCA63414310ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonium Bisulfite
CAS 10192-30-0
Molecular Formula H5NO3S
Quantity 10 mL
Synonym Ammonium Acid Sulfite, Ammonium Bisulfite, Ammonium Bisulfite (NH4HSO3), Ammonium Hydrogen Sulfite, Ammonium Monosulfite, Ammonium Sulfite (NH4HSO3), Monoammonium Sulfite, Solfoliquid 25, TC 1000
InChI Formula InChI=1S/H3N.H2O3S/c;1-4(2)3/h1H3;(H2,1,2,3)
SMILES [NH4+].OS(=O)[O-]
IUPAC Name azanium;hydrogen sulfite
Molecular Weight (g/mol) 99.11
Formula Weight 98.999
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.