missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bisulfite - In water, TRC
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | Ammonium Bisulfite |
| CAS | 10192-30-0 |
| Molecular Formula | H5NO3S |
| Quantity | 5 mL |
| Synonym | Ammonium Acid Sulfite, Ammonium Bisulfite, Ammonium Bisulfite (NH4HSO3), Ammonium Hydrogen Sulfite, Ammonium Monosulfite, Ammonium Sulfite (NH4HSO3), Monoammonium Sulfite, Solfoliquid 25, TC 1000 |
| InChI Formula | InChI=1S/H3N.H2O3S/c;1-4(2)3/h1H3;(H2,1,2,3) |
| SMILES | [NH4+].OS(=O)[O-] |
| IUPAC Name | azanium;hydrogen sulfite |
| Molecular Weight (g/mol) | 99.11 |
| Formula Weight | 98.999 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction