missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bisulfate, For decomposition, ≥99.0% (T), Honeywell™

Product Code. 15640920 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Poly bottle
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15640920 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15640920 Supplier Honeywell Research Chemicals Supplier No. 09849250G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 350°C
Melting Point 121°C to 145°C
CAS 7803-63-6
Color Colorless
pH 1 (100g/L at 20°C)
Physical Form Crystalline
Quantity 250 g
Molecular Formula H5NO4S
Linear Formula (NH4)HSO4
MDL Number MFCD00051073
UN Number UN2506
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
Solubility Information Soluble in water, 0.5g in 10mL
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
SMILES [NH4+].OS([O-])(=O)=O
IUPAC Name ammonium hydrogen sulfate
Molecular Weight (g/mol) 115.10
PubChem CID 24655
Formula Weight 115.11g/mol
Percent Purity ≥99.0% (T)
Packaging Poly bottle
Chemical Name or Material Ammonium Bisulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.